Drukuj

Anti-AIF1 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303093EA 2750 PLN
ORIGTA303093
Anti-AIF1 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3?g/ml; Immunochemistry: 2-4?g/ml

Type: Primary
Antigen: AIF1 (allograft inflammatory factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR