Drukuj

Anti-CDX2 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303095EA 2750 PLN
ORIGTA303095
Anti-CDX2 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:4000 Western Blot: 1-3µg/ml

Type: Primary
Antigen: CDX2 (caudal type homeobox 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR