Drukuj

Anti-SLC24A5 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303110EA 2750 PLN
ORIGTA303110
Anti-SLC24A5 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:2000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: SLC24A5 (solute carrier family 24 (sodium/potassium/calcium exchanger), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR