Drukuj

Anti-MYO9B Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303112EA 2750 PLN
ORIGTA303112
Anti-MYO9B Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:2000; Western Blot: 2-6µg/ml

Type: Primary
Antigen: MYO9B (myosin IXB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR