Drukuj

Anti-GPX4 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303125EA 2750 PLN
ORIGTA303125
Anti-GPX4 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:16000; Western Blot: 01-03µg/ml; Immunochemistry: 03µg/ml

Type: Primary
Antigen: GPX4 (glutathione peroxidase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR