Drukuj

Anti-GPX7 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303126EA 2750 PLN
ORIGTA303126
Anti-GPX7 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: GPX7 (glutathione peroxidase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR