Drukuj

Anti-VHL Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303127EA 2750 PLN
ORIGTA303127
Anti-VHL Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:128000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: VHL (von Hippel-Lindau tumor suppressor, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR