Drukuj

Anti-DCDC2 Goat polyclonal antibody

Dostawca: OriGene
ORIGTA303154EA 2750 PLN
ORIGTA303154
Anti-DCDC2 Goat polyclonal antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:32000 Western Blot: 1-3µg/ml

Type: Primary
Antigen: DCDC2 (doublecortin domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR