Drukuj

Anti-SLC7A11 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303158EA 2750 PLN
ORIGTA303158
Anti-SLC7A11 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:128000; Western Blot: 002-006µg/ml

Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR