Drukuj

Anti-CCM2 Goat polyclonal antibody

Dostawca: OriGene
ORIGTA303163EA 2750 PLN
ORIGTA303163
Anti-CCM2 Goat polyclonal antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:128000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: CCM2 (cerebral cavernous malformation 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR