Drukuj

Anti-GALR1 Goat polyclonal antibody

Dostawca: OriGene
ORIGTA303178EA 2750 PLN
ORIGTA303178
Anti-GALR1 Goat polyclonal antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: GALR1 (galanin receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR