Drukuj

Anti-AIRE Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303185EA 2750 PLN
ORIGTA303185
Anti-AIRE Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR