Drukuj

Anti-TIA1 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303222EA 2750 PLN
ORIGTA303222
Anti-TIA1 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: TIA1 (TIA1 cytotoxic granule-associated RNA binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR