Drukuj

Anti-DPP4 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303245EA 2750 PLN
ORIGTA303245
Anti-DPP4 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:64000 Western Blot: 03-1µg/ml

Type: Primary
Antigen: DPP4 (dipeptidyl-peptidase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR