Drukuj

Anti-DDX19B Goat polyclonal antibody

Dostawca: OriGene
ORIGTA303256EA 2750 PLN
ORIGTA303256
Anti-DDX19B Goat polyclonal antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:4000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: DDX19B (DEAD (Asp-Glu-Ala-Asp) box polypeptide 19B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR