Drukuj

Anti-GRIK1 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303274EA 2750 PLN
ORIGTA303274
Anti-GRIK1 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:8000 Western Blot: 1-3µg/ml

Type: Primary
Antigen: GRIK1 (glutamate receptor, ionotropic, kainate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR