Drukuj

Anti-ADRB3 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303277EA 2760 PLN
ORIGTA303277
Anti-ADRB3 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:128000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: ADRB3 (adrenoceptor beta 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR