Drukuj

Anti-B3GNT2 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303284EA 2750 PLN
ORIGTA303284
Anti-B3GNT2 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 32000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: B3GNT2 (UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR