Drukuj

Anti-SLC12A6 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303292EA 2750 PLN
ORIGTA303292
Anti-SLC12A6 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:16000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: SLC12A6 (solute carrier family 12 (potassium/chloride transporter), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR