Drukuj

Anti-PTGER3 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303309EA 2750 PLN
ORIGTA303309
Anti-PTGER3 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:2000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: PTGER3 (prostaglandin E receptor 3 (subtype EP3))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR