Drukuj

Anti-GOT2 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303325EA 2750 PLN
ORIGTA303325
Anti-GOT2 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:32000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: GOT2 (glutamic-oxaloacetic transaminase 2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR