Drukuj

Anti-PTPRN Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303345EA 2750 PLN
ORIGTA303345
Anti-PTPRN Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:16000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: PTPRN (protein tyrosine phosphatase, receptor type, N)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR