Drukuj

Anti-IGFBP4 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303356EA 2750 PLN
ORIGTA303356
Anti-IGFBP4 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: IGFBP4 (insulin-like growth factor binding protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR