Drukuj

Anti-SERPINF2 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303369EA 2750 PLN
ORIGTA303369
Anti-SERPINF2 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: SERPINF2 (serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR