Drukuj

Anti-TRPV2 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303371EA 2750 PLN
ORIGTA303371
Anti-TRPV2 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: TRPV2 (transient receptor potential cation channel, subfamily V, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR