Drukuj

Anti-APOF Goat antibody

Dostawca: OriGene
ORIGTA305632EA 2750 PLN
ORIGTA305632
Anti-APOF Goat antibody
Przeciwciała
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: APOF (apolipoprotein F)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR