Drukuj

Anti-P2RX7 Goat Antibody

Dostawca: OriGene
ORIGTA305642EA 2760 PLN
ORIGTA305642
Anti-P2RX7 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: P2RX7 (purinergic receptor P2X, ligand-gated ion channel, 7)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR