Drukuj

Anti-NKX2-5 Goat Antibody

Dostawca: OriGene
ORIGTA305653EA 2750 PLN
ORIGTA305653
Anti-NKX2-5 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: NKX2-5 (NK2 homeobox 5)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR