Drukuj

Anti-TMPRSS2 Goat Antibody

Dostawca: OriGene
ORIGTA305658EA 2750 PLN
ORIGTA305658
Anti-TMPRSS2 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: TMPRSS2 (transmembrane protease, serine 2)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR