Drukuj

Anti-MTNR1A Goat Antibody

Dostawca: OriGene
ORIGTA305660EA 2750 PLN
ORIGTA305660
Anti-MTNR1A Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: MTNR1A (melatonin receptor 1A)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR