Drukuj

Anti-TFB1M Goat Antibody

Dostawca: OriGene
ORIGTA305677EA 2750 PLN
ORIGTA305677
Anti-TFB1M Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: TFB1M (transcription factor B1, mitochondrial)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR