Drukuj

Anti-APOL2 Goat antibody

Dostawca: OriGene
ORIGTA305689EA 2750 PLN
ORIGTA305689
Anti-APOL2 Goat antibody
Przeciwciała
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: APOL2 (apolipoprotein L, 2)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR