Drukuj

Anti-COL4A3BP Goat Antibody

Dostawca: OriGene
ORIGTA305690EA 2750 PLN
ORIGTA305690
Anti-COL4A3BP Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: COL4A3BP (collagen, type IV, alpha 3 (Goodpasture antigen) binding protein)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR