Drukuj

Anti-DGAT2 Goat antibody

Dostawca: OriGene
ORIGTA305692EA 2750 PLN
ORIGTA305692
Anti-DGAT2 Goat antibody
Przeciwciała
Recommended Dilutions: Western Blot: 03-1?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: DGAT2 (diacylglycerol O-acyltransferase 2)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR