Drukuj

Anti-KCNQ1 Goat Antibody

Dostawca: OriGene
ORIGTA305704EA 2750 PLN
ORIGTA305704
Anti-KCNQ1 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: KCNQ1 (potassium voltage-gated channel, KQT-like subfamily, member 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR