Drukuj

Anti-MS4A1 Goat Antibody

Dostawca: OriGene
ORIGTA305713EA 2750 PLN
ORIGTA305713
Anti-MS4A1 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 0.03-0.1?g/ml

Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR