Drukuj

Anti-APOA1BP Goat Antibody

Dostawca: OriGene
ORIGTA305721EA 2750 PLN
ORIGTA305721
Anti-APOA1BP Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: APOA1BP (apolipoprotein A-I binding protein)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR