Drukuj

Anti-APOBEC3G Goat antibody

Dostawca: OriGene
ORIGTA305732EA 2750 PLN
ORIGTA305732
Anti-APOBEC3G Goat antibody
Przeciwciała
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: APOBEC3G (apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3G)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR