Drukuj

Anti-EVL Goat antibody

Dostawca: OriGene
ORIGTA305743EA 2760 PLN
ORIGTA305743
Anti-EVL Goat antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: EVL (Enah/Vasp-like)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR