Drukuj

Anti-TRIM2 Goat Antibody

Dostawca: OriGene
ORIGTA305745EA 2750 PLN
ORIGTA305745
Anti-TRIM2 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 05-2?g/ml; Immunochemistry: 6-12?g/ml

Type: Primary
Antigen: TRIM2 (tripartite motif containing 2)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR