Drukuj

Anti-PRDX1 Goat Antibody

Dostawca: OriGene
ORIGTA305773EA 2750 PLN
ORIGTA305773
Anti-PRDX1 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 001-003?g/ml

Type: Primary
Antigen: PRDX1 (peroxiredoxin 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR