Drukuj

Anti-SCGB2A2 Goat Antibody

Dostawca: OriGene
ORIGTA305775EA 2750 PLN
ORIGTA305775
Anti-SCGB2A2 Goat Antibody
Przeciwciała
Recommended Dilutions: Immunochemistry: 4-6?g/ml

Type: Primary
Antigen: SCGB2A2 (secretoglobin, family 2A, member 2)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR