Drukuj

Anti-HMOX2 Goat Antibody

Dostawca: OriGene
ORIGTA305779EA 2760 PLN
ORIGTA305779
Anti-HMOX2 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 0.3-1?g/ml; Immunochemistry: 2-4?g/ml

Type: Primary
Antigen: HMOX2 (heme oxygenase (decycling) 2)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR