Drukuj

Anti-CCT3 Goat Antibody

Dostawca: OriGene
ORIGTA305794EA 2750 PLN
ORIGTA305794
Anti-CCT3 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 001-003?g/ml

Type: Primary
Antigen: CCT3 (chaperonin containing TCP1, subunit 3 (gamma))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR