Drukuj

Anti-MAGOH Goat Antibody

Dostawca: OriGene
ORIGTA305811EA 2750 PLN
ORIGTA305811
Anti-MAGOH Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: MAGOH (mago-nashi homolog, proliferation-associated (Drosophila))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR