Drukuj

Anti-TNKS2 Goat Antibody

Dostawca: OriGene
ORIGTA305844EA 2750 PLN
ORIGTA305844
Anti-TNKS2 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: TNKS2 (tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR