Drukuj

Anti-ABCD3 Goat Antibody

Dostawca: OriGene
ORIGTA305866EA 2750 PLN
ORIGTA305866
Anti-ABCD3 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 02-06 ?g/ml

Type: Primary
Antigen: ABCD3 (ATP-binding cassette, sub-family D (ALD), member 3)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR