Drukuj

Anti-LRP4 Goat Antibody

Dostawca: OriGene
ORIGTA305882EA 2750 PLN
ORIGTA305882
Anti-LRP4 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: LRP4 (low density lipoprotein receptor-related protein 4)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR