Drukuj

Anti-SOCS7 Goat Antibody

Dostawca: OriGene
ORIGTA305889EA 2750 PLN
ORIGTA305889
Anti-SOCS7 Goat Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: SOCS7 (suppressor of cytokine signaling 7)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR