Drukuj

Anti-DFFB Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA305971EA 3010 PLN
ORIGTA305971
Anti-DFFB Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: DFFB (DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR